Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11014R-A680
BOSSBS-11014R-A680EA 890 EUR
BOSSBS-11014R-A680
Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ILVBL is a 632 amino acid single-pass membrane protein that belongs to the TPP enzyme family. Expressed in the majority of tissues, ILVBL has the highest level of expression in heart, pancreas and placenta. ILVBL is highly homologous to several bacterial enzymes, including the B isozyme of the large catalytic subunit of E. coli acetohydroxy-acid synthase (AHAS) and the oxalyl-coA decarboxylase of O. formigenes, that utilize thiamine pyrophosphate as a cofactor. ILVBL binds one magnesium ion and one thiamine pyrophosphate per subunit, and may catalyse the initial step in branched-chain amino acid biosynthesis. The gene encoding ILVBL maps to human chromosome 19p13.12 and mouse chromosome 10 C1.

Type: Primary
Antigen: ILVBL
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/632
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR