Anti-GALNT14 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11018R
BOSSBS-11018REA 704 EUR
BOSSBS-11018R
Anti-GALNT14 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a Golgi protein which is a member of the polypeptide N-acetylgalactosaminyltransferase (ppGalNAc-Ts) protein family. These enzymes catalyze the transfer of N-acetyl-D-galactosamine (GalNAc) to the hydroxyl groups on serines and threonines in target peptides. The encoded protein has been shown to transfer GalNAc to large proteins like mucins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 2011].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GALNT14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR