Anti-GALNT14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11018R-A488
BOSSBS-11018R-A488EA 876 EUR
BOSSBS-11018R-A488
Anti-GALNT14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This gene encodes a Golgi protein which is a member of the polypeptide N-acetylgalactosaminyltransferase (ppGalNAc-Ts) protein family. These enzymes catalyze the transfer of N-acetyl-D-galactosamine (GalNAc) to the hydroxyl groups on serines and threonines in target peptides. The encoded protein has been shown to transfer GalNAc to large proteins like mucins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALNT14
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR