Anti-GALNT14 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-11018R-A680
BOSSBS-11018R-A680EA 874 EUR
BOSSBS-11018R-A680
Anti-GALNT14 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes a Golgi protein which is a member of the polypeptide N-acetylgalactosaminyltransferase (ppGalNAc-Ts) protein family. These enzymes catalyse the transfer of N-acetyl-D-galactosamine (GalNAc) to the hydroxyl groups on serines and threonines in target peptides. The encoded protein has been shown to transfer GalNAc to large proteins like mucins. Multiple transcript variants encoding different isoforms have been found for this gene.
Type: Primary
Antigen: GALNT14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/552
Host: Rabbit
Isotype: IgG
Reactivity: Human