Anti-GAN Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-11025R-CY7
BOSSBS-11025R-CY7EA 893 EUR
BOSSBS-11025R-CY7
Anti-GAN Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Gigaxonin, also refered to as giant axonal neuropathy, GAN1, or KLHL16, controls protein degradation and is essential for neuronal function and survival. Gigaxonin is a member of the cytoskeletal BTB/kelch repeat family and influences cytoskeletal organization and dynamics, playing a large role in neurofilament architecture. The amino terminal BTB domain of gigaxonin binds to the ubiquitin-activating enzyme E1, while the carboxy-terminal kelch repeat domain interacts directly with the light chain of microtubule-associated protein 1B (MAP1B), and tags it for degredation. Overexpression of MAP1B may lead to neuronal cell death, whereas a reduction of MAP1B significantly improves the survival rate of neurons. Mutations in the Gigaxonin gene result in human giant axonal neuropathy (GAN), an autosomal recessive neurodegenerative disorder characterized by axonal degeneration caused by cytoskeletal abnormalities, including accumulated intermediate filaments.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gigaxonin
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR