Anti-MAP1B Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11028R
BOSSBS-11028REA 725 EUR
BOSSBS-11028R
Anti-MAP1B Rabbit Polyclonal Antibody
Antikörper
Facilitates tyrosination of alpha-tubulin in neuronal microtubules (By similarity). Phosphorylated MAP1B may play a role in the cytoskeletal changes that accompany neurite extension. Possibly MAP1B binds to at least two tubulin subunits in the polymer, and this bridging of subunits might be involved in nucleating microtubule polymerization and in stabilizing microtubules. Acts as a positive cofactor in DAPK1-mediated autophagic vesicle formation and membrane blebbing.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MAP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR