Anti-TNR/Tenascin R/TENR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11068R-A750
BOSSBS-11068R-A750EA 890 EUR
BOSSBS-11068R-A750
Anti-TNR/Tenascin R/TENR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The Tenascin family of extracellular matrix proteins includes Tenascin (also designated cytotactin or Tenascin-C), Tenascin-R (also designated Restrictin or Janusin) and Tenascin-X. Tenascin proteins function as substrate-adhesion molecules (SAMs) and are involved in regulating numerous developmental processes, such as morphogenetic cell migration and organogenesis. The Tenascin family proteins arise from various splicing events in the region of coding for FNIII repeats. Tenascin and Tenascin-X are expressed in several tissues during embryogenesis, and in adult tissues undergoing active remodel-ing such as healing wounds and tumours. Tenascin-R (TN-R) is expressed on the surface of neurons and glial cells.

Type: Primary
Antigen: TNR/Tenascin R/TENR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1021-1120/1358
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR