Anti-TENR Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-11068R-CY3
BOSSBS-11068R-CY3EA 900 EUR
BOSSBS-11068R-CY3
Anti-TENR Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The Tenascin family of extracellular matrix proteins includes Tenascin (also designated cytotactin or Tenascin-C), Tenascin-R (also designated Restrictin or Janusin) and Tenascin-X. Tenascin proteins function as substrate-adhesion molecules (SAMs) and are involved in regulating numerous developmental processes, such as morphogenetic cell migration and organogenesis. The Tenascin family proteins arise from various splicing events in the region of coding for FNIII repeats. Tenascin and Tenascin-X are expressed in several tissues during embryogenesis, and in adult tissues undergoing activeMore Product Information