Anti-TENR Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-11068R-CY5
BOSSBS-11068R-CY5EA 893 EUR
BOSSBS-11068R-CY5
Anti-TENR Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Tenascin family of extracellular matrix proteins includes Tenascin (also designated cytotactin or Tenascin-C), Tenascin-R (also designated Restrictin or Janusin) and Tenascin-X. Tenascin proteins function as substrate-adhesion molecules (SAMs) and are involved in regulating numerous developmental processes, such as morphogenetic cell migration and organogenesis. The Tenascin family proteins arise from various splicing events in the region of coding for FNIII repeats. Tenascin and Tenascin-X are expressed in several tissues during embryogenesis, and in adult tissues undergoing active remodel-ing such as healing wounds and tumors. Tenascin-R (TN-R) is expressed on the surface of neurons and glial cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Tenascin R
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat