Anti-4-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1111R-A680
BOSSBS-1111R-A680EA 874 EUR
BOSSBS-1111R-A680
Anti-4-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). Forms a trimeric complex with SOX2 on DNA and controls the expression of a number of genes involved in embryonic development such as YES1, FGF4, UTF1 and ZFP206. Critical for early embryogenesis and for embryonic stem cell pluripotency.

Type: Primary
Antigen: 4-Oct
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/360
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR