Anti-4-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-1111R-A750
BOSSBS-1111R-A750EA 874 EUR
BOSSBS-1111R-A750
Anti-4-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). Forms a trimeric complex with SOX2 on DNA and controls the expression of a number of genes involved in embryonic development such as YES1, FGF4, UTF1 and ZFP206. Critical for early embryogenesis and for embryonic stem cell pluripotency.
Type: Primary
Antigen: 4-Oct
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/360
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others