Anti-POU5F1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1111R-CY5
BOSSBS-1111R-CY5EA 876 EUR
BOSSBS-1111R-CY5
Anti-POU5F1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). Forms a trimeric complex with SOX2 on DNA and controls the expression of a number of genes involved in embryonic development such as YES1, FGF4, UTF1 and ZFP206. Critical for early embryogenesis and for embryonic stem cell pluripotency.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat