Anti-MAdCAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11179R-A750
BOSSBS-11179R-A750EA 890 EUR
BOSSBS-11179R-A750
Anti-MAdCAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Cell adhesion leukocyte receptor expressed by mucosal venules, helps to direct lymphocyte traffic into mucosal tissues including the Peyer patches and the intestinal lamina propria. It can bind both integrin alpha-4/beta-7 and L-selectin, regulating both the passage and retention of leukocytes. Isoform 2, lacking the mucin-like domain, may be specialised in supporting integrin alpha-4/beta-7-dependent adhesion strengthening, independent of L-selectin binding.

Type: Primary
Antigen: MAdCAM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 85-180/382
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR