Anti-ABCA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-11180R-A647
BOSSBS-11180R-A647EA 893 EUR
BOSSBS-11180R-A647
Anti-ABCA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of widely-expressed proteins that use ATP hydrolysis to catalyze the transport of various molecules across extracellular and intracellular membranes. Eukaryotic ABC transporters are largely responsible for trafficking hydrophobic compounds either within the cell as part of a metabolic process, outside the cell for transport to other organs, or for secretion from the body. The cholesterol-responsive transporter, ABCA7, maps to human chromosome 19 and mouse chromosome 10 and has been reported as a candidateMore Product Information