Anti-Neuroligin 3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11181R-A750
BOSSBS-11181R-A750EA 890 EUR
BOSSBS-11181R-A750
Anti-Neuroligin 3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.

Type: Primary
Antigen: Neuroligin 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 601-700/848
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR