Anti-MOBP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11184R-A680
BOSSBS-11184R-A680EA 890 EUR
BOSSBS-11184R-A680
Anti-MOBP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
May play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.

Type: Primary
Antigen: MOBP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 7-90/183
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR