Anti-ARHGAP26 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11248R-CY5
BOSSBS-11248R-CY5EA 900 EUR
BOSSBS-11248R-CY5
Anti-ARHGAP26 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Cellular signaling by G-proteins is down-regulated by GTPase-activating proteins (GAPs), which increase the rate of GTP hydroylsis. The GTPase regulator associated with focal adhesion kinase (GRAF) has GAP activity toward Rho A and Cdc42, but not Rac1. GRAF is ubiquitously expressed with high levels in heart and brain. Expression of GRAF causes clearing of stress fibers and formation of long actin based filopodial-like extensions. Fusion of MLL with GRAF, MLL/GRAF, is included in a rare genetic subgroup of acute myeloid leukemia (AML) cases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARHGAP26
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR