Anti-PIST/GOPC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-11253R-A750
BOSSBS-11253R-A750EA 890 EUR
BOSSBS-11253R-A750
Anti-PIST/GOPC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PIST (PDZ protein interacting specifically with TC10), also known as GOPC (golgi associated PDZ and coiled-coil motif containing), CAL or FIG, is a 462 amino acid protein that localizes to the cytoplasm, as well as to the membrane of the golgi apparatus and to the cell junction. Expressed ubiquitously and containing one PDZ (DHR) domain, PIST functions as a homooligomer that interacts with a variety of proteins and plays a role in intracellular protein trafficking and degradation. Additionally, PIST is thought to regulate ionic currents via membrane channel modification and may also play a role in autophagy. Chromosomal aberrations in the gene encoding PIST are found in glioblastoma multiform (GBM), a common and aggressive form of brain tumour, suggesting a role for mutated PIST in carcinogenesis. Three isoforms of PIST exist due to alternative splicing events.
Type: Primary
Antigen: PIST/GOPC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/462
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen