Anti-5-HT Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-1126R-A647
BOSSBS-1126R-A647EA 876 EUR
BOSSBS-1126R-A647
Anti-5-HT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Serotonin (pronounced) (5-hydroxytryptamine, or 5-HT) is a monoamine neurotransmitter synthesized in serotonergic neurons in the central nervous system (CNS) and enterochromaffin cells in the gastrointestinal tract of animals including humans. Serotonin is also found in many mushrooms and plants, including fruits and vegetables.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 5-HT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR