Anti-LCN1/lipocalin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11286R-A680
BOSSBS-11286R-A680EA 890 EUR
BOSSBS-11286R-A680
Anti-LCN1/lipocalin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Lipocalin-1 is a secretory protein that is highly expressed in fluids covering epithelial surfaces such as tears and respiratory secretions. This major lipid-binding protein in tears is also called tear lipocalin (TL) and von Ebner's gland protein (VEG), as it is also a major secretion of these lingual salivary glands. In addition to lacrimal glands and lingual glands, Lipocalin-1 is secreted by nasal mucosal glands, secretory glands of the tracheobronchial tract, sweat glands, mammary glands, adrenal gland, prostate, thymus, testis and corticotrophs of the pituitary gland. Specifically, Lipocalin-1 functions to stabilize the lipid film of human tear fluid by removing harmful lipids from the human corneal surface and delivering them to the aqueous phase of tears. Lipocalin-1 may also function as a transporter of hydrophobic molecules such as bitter substances on the tongue.

Type: Primary
Antigen: LCN1/lipocalin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-176/176
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR