Anti-GBP2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-11289R-CY7
BOSSBS-11289R-CY7EA 1350 EUR
BOSSBS-11289R-CY7
Anti-GBP2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Hydrolyzes GTP to GMP in two consecutive cleavage reactions. Exhibits antiviral activity against influenza virus. Promote oxidative killing and deliver antimicrobial peptides to autophagolysosomes, providing broad host protection against different pathogen classes (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GBP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat