Anti-PTP zeta Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11327R-A680
BOSSBS-11327R-A680EA 890 EUR
BOSSBS-11327R-A680
Anti-PTP zeta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important Signalling cascades (By similarity).

Type: Primary
Antigen: PTP zeta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-520/2315
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR