Anti-GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-11381R-A488
BOSSBS-11381R-A488EA 893 EUR
BOSSBS-11381R-A488
Anti-GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Contributes to degradation of proteins cross-linked by transglutaminases. Degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. Catalyzes the formation of 5-oxoproline from L-gamma-glutamyl-L-epsilon-lysine. Inactive with L-gamma-glutamyl-alpha-amino acid substrates such as L-gamma-glutamyl-L-alpha-cysteine and L-gamma-glutamyl-L-alpha-alanine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: A2LD1/GGACT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat