Anti-GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-11381R-A555
BOSSBS-11381R-A555EA 893 EUR
BOSSBS-11381R-A555
Anti-GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Contributes to degradation of proteins cross-linked by transglutaminases. Degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. Catalyzes the formation of 5-oxoproline from L-gamma-glutamyl-L-epsilon-lysine. Inactive with L-gamma-glutamyl-alpha-amino acid substrates such as L-gamma-glutamyl-L-alpha-cysteine and L-gamma-glutamyl-L-alpha-alanine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: A2LD1/GGACT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR