Anti-A2LD1/GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11381R-A680
BOSSBS-11381R-A680EA 890 EUR
BOSSBS-11381R-A680
Anti-A2LD1/GGACT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Contributes to degradation of proteins cross-linked by transglutaminases. Degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. catalyses the formation of 5-oxoproline from L-gamma-glutamyl-L-epsilon-lysine. Inactive with L-gamma-glutamyl-alpha-amino acid substrates such as L-gamma-glutamyl-L-alpha-cysteine and L-gamma-glutamyl-L-alpha-alanine.

Type: Primary
Antigen: A2LD1/GGACT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-50/153
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR