Anti-ZNF318 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-11414R-FITC
BOSSBS-11414R-FITCEA 900 EUR
BOSSBS-11414R-FITC
Anti-ZNF318 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ZNF318 is a 2279 amino acid endocrine regulatory protein that localizes to the nucleus. Highly expressed in testis, ovaries and kidneys, ZNF318 is a co-repressor of androgen receptor (AR)-mediated transcriptional activation and is thought to regulate transcription during spermatogenesis. ZNF318 interacts with the N-terminal domain of AR and contains two matrin-type zinc fingers. Two isoforms of ZNF318, designated TZF and TZF-L, are produced due to alternative splicing events. Each of these splice variants are thought to have unique roles in transcriptional regulation. While the TZF isoform functions as a repressor of AR-mediated transcriptional activation, the TZF-L isoform is thought to enhance AR-mediated transcriptional activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF318
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR