Anti-SLC16A2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11434R-CY5
BOSSBS-11434R-CY5EA 893 EUR
BOSSBS-11434R-CY5
Anti-SLC16A2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Very active and specific thyroid hormone transporter. Stimulates cellular uptake of thyroxine (T4), triiodothyronine (T3), reverse triiodothyronine (rT3) and diidothyronine. Does not transport Leu, Phe, Trp or Tyr (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC16A2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR