Anti-NG Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-11435R-CY3
BOSSBS-11435R-CY3EA 900 EUR
BOSSBS-11435R-CY3
Anti-NG Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Acts as a "third messenger" substrate of protein kinase C-mediated molecular cascades during synaptic development and remodeling. Binds to calmodulin in the absence of calcium (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NG/Neurogranin
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse