Anti-TRIP13/16E1BP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11438R-A680
BOSSBS-11438R-A680EA 890 EUR
BOSSBS-11438R-A680
Anti-TRIP13/16E1BP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumour suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.

Type: Primary
Antigen: TRIP13/16E1BP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/432
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR