Anti-AANAT Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-11448R-A350
BOSSBS-11448R-A350EA 921 EUR
BOSSBS-11448R-A350
Anti-AANAT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Arylalkylamine N-acetyltransferase belongs to the superfamily of acetyltransferases. It is the penultimate enzyme in melatonin synthesis and controls the night/day rhythm in melatonin production in the vertebrate pineal gland. Melatonin is essential for seasonal reproduction, modulates the function of the circadian clock in the suprachiasmatic nucleus, and influences activity and sleep. This enzyme is rapidly inactivated when animals are exposed to light at night. This protein is 80% identical to sheep and rat AA-NAT. Arylalkylamine N-acetyltransferase may contribute a multifactorial genetic diseases such as altered behavior in sleep/wake cycle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AANAT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR