Anti-Ephexin-1/EPHEXIN Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11458R-A488
BOSSBS-11458R-A488EA 893 EUR
BOSSBS-11458R-A488
Anti-Ephexin-1/EPHEXIN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
EPHEXIN is a 710 amino acid protein that localizes to both the membrane and the cytoplasm and contains one SH3 domain, one PH domain and one DH domain. Expressed at high levels in brain and present at lower levels in lung tissue, EPHEXIN interacts with EphA4 and functions as a guanine nucleotide exchange factor (GEF) that is capable of activating Rho A, Rac 1 and Cdc42 and is thought to play a role in axon guidance and growth cone collapse. EPHEXIN is subject to Src-dependent phosphorylation, an event that increases the GEF activity of EPHEXIN toward Rho A. Human EPHEXIN, which exists as multiple alternatively spliced isoforms, shares a high degree of sequence homology with its mouse counterpart, suggesting a conserved role between species.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ephexin-1/EPHEXIN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR