Anti-SPEF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11488R-A488
BOSSBS-11488R-A488EA 893 EUR
BOSSBS-11488R-A488
Anti-SPEF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Flagella and cillia are both membrane-bound projections from the cell surface that beat in distinctive patterns. Cilia are shorter and usually more profuse than flagella and contain a microtubule cytoskeleton, the ciliary axoneme, surrounded by a ciliary membrane. The ciliary membranes of all cilia hold specific receptors and ion channel proteins that initiate signaling pathways that regulate motility and/or link mechanical or chemical stimuli to intracellular transduction cascades regulating differentiation, migration and cell growth during development and in adulthood. KPL2, also known as SPEF2 (sperm flagellar 2), is a 1,822 amino acid protein that contains a calponin homology domain, three nuclear localization signals, a consensus P-loop and a proline-rich region. Required for correct axoneme develoment, KPL2 is predominantly expressed in cells with cilia or flagella. Four isoforms of KPL2 exists as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPEF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR