Anti-KPL2/Sperm Flagellar 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-11488R-A750
BOSSBS-11488R-A750EA 890 EUR
BOSSBS-11488R-A750
Anti-KPL2/Sperm Flagellar 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Flagella and cilia are both membrane-bound projections from the cell surface that beat in distinctive patterns. Cilia are shorter and usually more profuse than flagella and contain a microtubule cytoskeleton, the ciliary axoneme, surrounded by a ciliary membrane. The ciliary membranes of all cilia hold specific receptors and ion channel proteins that initiate Signalling pathways that regulate motility and/or link mechanical or chemical stimuli to intracellular transduction cascades regulating differentiation, migration and cell growth during development and in adulthood. KPL2, also known asMore Product Information