Anti-TNRC6B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-11489R-HRP
BOSSBS-11489R-HRPEA 900 EUR
BOSSBS-11489R-HRP
Anti-TNRC6B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
TNRC6B is a 1,723 amino acid protein that exists as two alternatively spliced isoforms and is thought to be involved in mRNA cleavage events. Expressed ubiquitously, TNRC6B contains one glycine/tryptophan (GW)-rich N-terminal domain, one central glutamine-rich region and one C-terminal RNA recognition motif and is encoded by a gene that maps to human chromosome 22. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia. Additionally, translocations between chromosomes 9 and 22 may lead to the formation of the Philadelphia Chromosome and the subsequent production of the novel fusion protein BCR-Abl, a potent cell proliferation activator found in several types of leukemias.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TNRC6B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR