Anti-HS1BP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-11568R-A647
BOSSBS-11568R-A647EA 893 EUR
BOSSBS-11568R-A647
Anti-HS1BP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
HS1BP3 is a 392 amino acid protein that contains one PX (phox homology) domain, a leucine zipper, immunoreceptor tyrosine-based inhibitory motif-like motifs and multiple proline-rich regions. Expressed primarily in brain, HS1BP3 is encoded by a gene mapping to human chromosome 2p24.1. The gene encoding HS1BP3 is frequently mutated in familial essential tremor, a disorder characterized by kinetic tremor the the hands, voice or head, though there is no correlation to Parkinson disease. HS1BP3 interacts with HAX-1’s SH3 domain, and may also play a role in the regulation of catecholamine and serotonin metabolism. Acting as a regulator of IL-2 signaling, HS1BP3 is likely involved in lymphocyte activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HS1BP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR