Anti-RAPGEF2/PDZ-GEF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11581R-A750
BOSSBS-11581R-A750EA 890 EUR
BOSSBS-11581R-A750
Anti-RAPGEF2/PDZ-GEF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PDZ-GEF1 is a 1499 amino acid cell membrane protein that functions as a guanine nucleotide exchange factor for Rap 1A, Rap 1B and Rap 2B GTPases. Expressed at highest levels in brain, PDZ-GEF1 is found at low levels in placenta, heart, lung and kidney, and undergoes post-translational phosphorylation following DNA damage. PDZ-GEF1 interacts with MAGI-2 and contains one Ras-GEF domain, a Ras-associating domain, one PDZ (DHR) domain, a single N-terminal Ras-GEF domain and a cyclic nucleotide-binding domain. The gene encoding PDZ-GEF1 maps to human chromosome 4q32.1.

Type: Primary
Antigen: RAPGEF2/PDZ-GEF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1499
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR