Anti-HOXC5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-11589R-A555
BOSSBS-11589R-A555EA 893 EUR
BOSSBS-11589R-A555
Anti-HOXC5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The Hox homeobox genes encode proteins that are transcriptional regulators with an established role in embryonic development. HoxC5 is a 222 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. Existing as multiple alternatively spliced isoforms, HoxC5 functions as a sequence-specific DNA-binding transcription factor that is part of a regulatory mechanism that provides cells with positional identities during development. The gene encoding HoxC5 maps to a cluster of Hox proteins on chromosome 12 that are essential for morphogenesis. Encoding over 1,100 genes within 132 million bases, chromosome 12 makes up about 4.5% of the human genome. A number of skeletal deformities are linked to chromosome 12, including hypochondrogenesis, achondrogenesis and Kniest dysplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HOXC5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR