Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11595R-A488
BOSSBS-11595R-A488EA 893 EUR
BOSSBS-11595R-A488
Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Three highly similar proteins termed NOMO1, NOMO2 and NOMO3, are encoded by a gene mapping to a region of duplication on the p arm of human chromosome 16. All three NOMO proteins share similar functions and have been difficult to characterize individually. NOMO1 (Nodal modulator 1), also known as PM5, is a 1,222 amino acid highly conserved single-pass type I membrane protein expressed in colon tumor tissue and normal colonic mucosa. NOMO proteins are novel antagonists of Nodal signaling which interact with Nicalin to form a Nicalin-NOMO complex, and are rapidly degraded or stabilized by Nicalin. NOMO proteins were once considered candidates for the development of pseudoxanthoma elasticum (PXE), a heritable disorder of connective tissue, as the NOMO genes are located in close proximity to the gene responsible for PXE development (MRP6).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOMO1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR