Anti-TWSG1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-11606R-A647
BOSSBS-11606R-A647EA 893 EUR
BOSSBS-11606R-A647
Anti-TWSG1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Twisted Gastrulation Protein 1(TWSG1) is a secreted BMP binding protein structurally related to the BMP antagonists Chordin and Noggin. TSG can inhibit BMP activity by binding directly to BMP proteins thereby preventing BMPs from binding to their receptors. It can act either as a BMP4 agonist or antagonist (depending on the specific biochemical environment) by binding to the BMP4/Chordin complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TWSG1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR