Anti-ZIC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-11610R-A488
BOSSBS-11610R-A488EA 893 EUR
BOSSBS-11610R-A488
Anti-ZIC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Zic2 is a C2H2 zinc finger transcription factor that influences forebrain development. Zic2 is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. Zic2 transcript is abundant in the dorsal neural tube/spinal cord, and in the hindbrain. A polyhistidine tract polymorphism in this gene may be associated with increased risk of neural tube defects. This gene is closely linked to a gene encoding zinc finger protein of the cerebellum 5, a related family member on chromosome 13.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZIC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat