Anti-TAS2R5 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11614R-CY5
BOSSBS-11614R-CY5EA 900 EUR
BOSSBS-11614R-CY5
Anti-TAS2R5 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This gene encodes a bitter taste receptor; bitter taste receptors are members of the G protein-coupled receptor superfamily and are specifically expressed by taste receptor cells of the tongue and palate epithelia. Each of these apparently intronless taste receptor genes encodes a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes on chromosome 7 and is genetically linked to loci that influence bitter perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAS2R5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR