Anti-TAS2R50 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11617R-A488
BOSSBS-11617R-A488EA 893 EUR
BOSSBS-11617R-A488
Anti-TAS2R50 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The sense of taste is essential for the survival of organisms. For example, the ability to identify sweet-tasting foods enables animals to seek out food with high nutritive value, whereas the ability to identify bitter substances enables them to avoid the ingestion of potentially harmful substances. A family of integral membrane proteins are involved in taste perception and include T1R, which is involved in sweet taste perception and T2R, which is involved in bitter taste perception. Both types of taste receptors couple to various G proteins to initiate signal transduction cascades. Specifically, T2R50 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAS2R50
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR