Anti-OR5P3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11628R
BOSSBS-11628REA 725 EUR
BOSSBS-11628R
Anti-OR5P3 Rabbit Polyclonal Antibody
Antikörper
Odorant receptor (Potential). May be involved in taste perception.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: OR5P3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR