Anti-LZIC Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11638R-A680
BOSSBS-11638R-A680EA 890 EUR
BOSSBS-11638R-A680
Anti-LZIC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
LZIC is a 190 amino acid protein that belongs to the CTNNBIP1 family. Ubiquitously expressed, LZIC is found at highest levels in kidney and is upregulated in a few gastric carcinomas. The gene encoding LZIC maps to human chromosome 1, which spans 260 million base pairs, contains over 3000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.

Type: Primary
Antigen: LZIC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/190
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR