Anti-DEFB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-1163R-CY5
BOSSBS-1163R-CY5EA 900 EUR
BOSSBS-1163R-CY5
Anti-DEFB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis. The mature form of Beta defensin 1 is 36 amino acids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR