Anti-BPTF Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11641R
BOSSBS-11641REA 725 EUR
BOSSBS-11641R
Anti-BPTF Rabbit Polyclonal Antibody
Antikörper
BPTF is a 2,907 amino acid protein encoded by the human gene BPTF. BPTF belongs to the PBTF family and contains one bromodomain, one DDT domain and two PHD-type zinc fingers. BPTF acts as a histone-binding component of NURF (nucleosome-remodeling factor). The NURF complex, which consists of SMARCA1, BPTF, RbAp46 and RbAp48, acts to catalyze ATP-dependent nucleosome sliding and facilitates transcription of chromatin. It specifically recognizes histone H3 tails trimethylated on 'Lys-4' (H3-K4Me3), which mark transcription start sites of virtually all active genes. BPTF may also help regulate transcription through direct binding to DNA or transcription factors.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: BPTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR