Anti-FISH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-11645R-A680
BOSSBS-11645R-A680EA 890 EUR
BOSSBS-11645R-A680
Anti-FISH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Fish, a potential Src substrate, is a broadly expressed adaptor protein containing five SH3 domains and a phox homology (PX) domain. The Src family of protein tyrosine kinases act in signal transduction pathways. Src kinases vary in expression but are strongly regulated in vivo; catalytic activity is repressed by interacting with the SH3 domain. In Src-transformed fibroblasts and in normal cells treated with certain growth factors fish is tyrosine-phosphorylated. Treatment of cells with cytochalasin D results in rapid tyrosine phosphorylation of fish, along with activation of Src. Fish is likely to be involved in tyrosine kinase Signalling and may have a role in cytoskeletal changes.
Type: Primary
Antigen: FISH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 261-360/1133
Host: Rabbit
Isotype: IgG
Reactivity: Rat