Anti-FISH Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11645R-A750
BOSSBS-11645R-A750EA 890 EUR
BOSSBS-11645R-A750
Anti-FISH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Fish, a potential Src substrate, is a broadly expressed adaptor protein containing five SH3 domains and a phox homology (PX) domain. The Src family of protein tyrosine kinases act in signal transduction pathways. Src kinases vary in expression but are strongly regulated in vivo; catalytic activity is repressed by interacting with the SH3 domain. In Src-transformed fibroblasts and in normal cells treated with certain growth factors fish is tyrosine-phosphorylated. Treatment of cells with cytochalasin D results in rapid tyrosine phosphorylation of fish, along with activation of Src. Fish is likely to be involved in tyrosine kinase Signalling and may have a role in cytoskeletal changes.

Type: Primary
Antigen: FISH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-360/1133
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR