Anti-FISH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-11645R-HRP
BOSSBS-11645R-HRPEA 900 EUR
BOSSBS-11645R-HRP
Anti-FISH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Fish, a potential Src substrate, is a broadly expressed adaptor protein containing five SH3 domains and a phox homology (PX) domain (1). The Src family of protein tyrosine kinases act in signal transduction pathways (2-4). Src kinases vary in expression but are strongly regulated in vivo; catalytic activity is repressed by interacting with the SH3 domain (5-7). In Src-transformed fibroblasts and in normal cells treated with certain growth factors fish is tyrosine-phosphorylated (1). Treatment of cells with cytochalasin D results in rapid tyrosine phosphorylation of fish, along with activation of Src (1). Fish is likely to be involved in tyrosine kinase signaling and may have a role in cytoskeletal changes (1).

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: FISH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR