Anti-BLMH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11654R-A680
BOSSBS-11654R-A680EA 890 EUR
BOSSBS-11654R-A680
Anti-BLMH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Bleomycin hydrolase (BMH) is a cytoplasmic cysteine peptidase that is highly conserved through evolution; however, the only known activity of the enzyme is metabolic inactivation of the glycopeptide bleomycin (BLM), an essential component of combination chemotherapy regimens for cancer. The protein contains the signature active site residues of the cysteine protease papain superfamily.

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 308-400/455
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR